3,3'-Dimethoxybenzil 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Bis(3-methoxyphenyl)ethane-1,2-dione, Bis(3-methoxyphenyl)ethanedione

L11564-03 L11564-06
CAAAAL11564-03EA 58.78 CAD
CAAAAL11564-03 CAAAAL11564-06
3,3'-Dimethoxybenzil 99+%
3,3'-Dimethoxybenzil

Purity: 99+% CAS No.: 40101-17-5 Molecular Formula: C16H14O4 Formula Weight: 270.29 Melting Point: 81-83° Beilstein Reference: 2382124

Formula: C₁₆H₁₄O₄
MW: 270.28 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00038221
CAS Number: 40101-17-5
EINECS: 254-793-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR