2-Ethyl-1-butene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11553-06
L11553-14
CAAAAL11553-06EA
103.65
CAD
CAAAAL11553-06
CAAAAL11553-14
2-Ethyl-1-butene 97%
2-Ethyl-1-butene
Purity: 97%
CAS No.: 760-21-4
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -26°(-14°F)
Refractive Index: 1.3960
Melting Point: -132°
Boiling Point: 63-64°
Beilstein Reference: 1697102
UN No.: 3295
MDL No.: MFCD00009341
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 65 Melting Pt: -131.5 Density: 0.689 Flash Pt: –26 °C (–14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009341 CAS Number: 760-21-4 EINECS: 212-078-5 UN: 3295 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...