2-Mercaptobenzyl alcohol tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

L11536-03 L11536-06 L11536-14
CAAAAL11536-03EA 66.2 CAD
CAAAAL11536-03 CAAAAL11536-06 CAAAAL11536-14
2-Mercaptobenzyl alcohol tech. 90%, Technical Grade
2-Mercaptobenzyl alcohol

Purity: 90% CAS No.: 4521-31-7 Molecular Formula: C7H8OS Formula Weight: 140.21 Flash Point: 110°(230°F) Refractive Index: 1.6230 Odor: Like rotten eggs Melting Point: 31-32° Boiling Point: 94-96°/0.2mm Beilstein Reference: 2206253 MDL No.: MFCD00014448

Formula: C₇H₈OS
Boiling Pt: 94…96 °C/0.2 mm
Melting Pt: 31…32 °C
Density: 1.210
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014448
CAS Number: 4521-31-7
UN: 3335
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR