(R)-(-)-2-Octanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11502-03
L11502-06
CAAAAL11502-03EA
82.48
CAD
CAAAAL11502-03
CAAAAL11502-06
(R)-(-)-2-Octanol 99%
(R)-(-)-2-Octanol
Purity: 99%
CAS No.: 5978-70-1
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 76°(169°F)
Refractive Index: 1.4260
Odor: Pleasant
Boiling Point: 179-180°
Beilstein Reference: 1719324
Merck Reference: 14,6752
Optical Rotation: -10° (c=1 in ethanol)
MDL No.: MFCD00064284
Formula:
C₈H₁₈O Boiling Pt: 179…180 °C Density: 0.819 Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064284 CAS Number: 5978-70-1 EINECS: 227-777-0 UN: 1986 ADR: 3,III Merck Index: 14,6752 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...