4-Bromobenzyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene
L11486-06
L11486-14
CAAAAL11486-06EA
80.11
CAD
CAAAAL11486-06
CAAAAL11486-14
4-Bromobenzyl chloride 97%
4-Bromobenzyl chloride
Purity: 97%
CAS No.: 589-17-3
Molecular Formula: C7H6BrCl
Formula Weight: 205.49
Flash Point:
Formula:
C₇H₆BrCl Boiling Pt: 106…107 °C/10 mm Melting Pt: 38…41 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040867 CAS Number: 589-17-3 EINECS: 209-638-6 UN: 3261 ADR: 8,III Merck Index: 14,1410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...