(-)-Dimethyl-D-tartrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: D(-)-Tartaric acid dimethyl ester, (2S,3S)-Dimethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Tartaric acid dimethyl ester, (S,S)-(-)-Dimethyl tartrate, (-)-Dimethyl D-tartrate

L11472-06 L11472-14
CAAAAL11472-06EA 85.32 CAD
CAAAAL11472-06 CAAAAL11472-14
(-)-Dimethyl-D-tartrate 99%
(-)-Dimethyl D-tartrate

Purity: 99% CAS No.: 13171-64-7 Molecular Formula: C6H10O6 Formula Weight: 178.14 Flash Point: 170°(338°F) Melting Point: 48-56° Boiling Point: 158°/11mm Beilstein Reference: 1726254 Optical Rotation: -21° (c=1 in water) MDL No.: MFCD00008445

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 158 °C/11 mm
Melting Pt: 48…56 °C
Density: 1.30
Flash Pt: >170 °C(338 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008445
CAS Number: 13171-64-7
EINECS: 236-118-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR