Benzoyl fluoride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11464-06
L11464-14
CAAAAL11464-06EA
58.78
CAD
CAAAAL11464-06
CAAAAL11464-14
Benzoyl fluoride 97%
Benzoyl fluoride
Purity: 97%
CAS No.: 455-32-3
Molecular Formula: C7H5FO
Formula Weight: 124.12
Flash Point: 48°(118°F)
Refractive Index: 1.4960
Odor: Pungent
Melting Point: ca -28°
Boiling Point: 159-161°
Beilstein Reference: 1904977
UN No.: 2920
MDL No.: MFCD00000364
Formula:
C₇H₅FO Boiling Pt: 159…161 °C Melting Pt: ∼ 28 °C Density: 1.140 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000364 CAS Number: 455-32-3 EINECS: 207-244-9 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...