4-Hydroxybutanone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-2-butanone, 4-Hydroxybutan-2-one

L11456-14 L11456-22
CAAAAL11456-14EA 88.64 CAD
CAAAAL11456-14 CAAAAL11456-22
4-Hydroxybutanone 95%
4-Hydroxybutanone

Purity: 95% CAS No.: 590-90-9 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 89°(192°F) Refractive Index: 1.4310 Boiling Point: 73-76°/12mm Beilstein Reference: 1737666 MDL No.: MFCD00059005

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 73…76 °C/12 mm
Density: 1.020
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059005
CAS Number: 590-90-9
EINECS: 209-693-6
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR