(3,5-Dimethoxyphenyl)acetonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile
L11443-03
L11443-06
CAAAAL11443-03EA
83.42
CAD
CAAAAL11443-03
CAAAAL11443-06
(3,5-Dimethoxyphenyl)acetonitrile 98+%
(3,5-Dimethoxyphenyl)acetonitrile
Purity: 98+%
CAS No.: 13388-75-5
Molecular Formula: C10H11NO2
Formula Weight: 177.20
Odor: Characteristic
Melting Point: 53-56°
Beilstein Reference: 2096481
MDL No.: MFCD00016395
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: 53…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016395 CAS Number: 13388-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...