3,3'-Diamino diphenyl sulfone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11400-14
L11400-22
CAAAAL11400-14EA
54.04
CAD
CAAAAL11400-14
CAAAAL11400-22
3,3'-Diamino diphenyl sulfone 98%
Purity: 98%
CAS No.: 599-61-1
Molecular Formula: C12H12N2O2S
Formula Weight: 248.30
Odor: Odorless
Melting Point: 171-174°
Beilstein Reference: 2215938
MDL No.: MFCD00007792
Formula:
C₁₂H₁₂N₂O₂S Melting Pt: 171…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007792 CAS Number: 599-61-1 EINECS: 209-967-5 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...