2-Bromo-3-methylthiophene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11393-06
L11393-14
CAAAAL11393-06EA
53.56
CAD
CAAAAL11393-06
CAAAAL11393-14
2-Bromo-3-methylthiophene 97%
2-Bromo-3-methylthiophene
Purity: 97%
CAS No.: 14282-76-9
Molecular Formula: C5H5BrS
Formula Weight: 177.07
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5710
Boiling Point: 173-176°
Beilstein Reference: 105719
UN No.: 2810
MDL No.: MFCD00059741
Formula:
C₅H₅BrS Boiling Pt: 173…176 °C Density: 1.580 Flash Pt: 68 °C(154 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00059741 CAS Number: 14282-76-9 EINECS: 238-175-2 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...