1-Phenylisatin 98%

Supplier: Thermo Scientific Chemicals
Danger

L11388-03 L11388-06
CAAAAL11388-03EA 63.36 CAD
CAAAAL11388-03 CAAAAL11388-06
1-Phenylisatin 98%
1-Phenylisatin

Purity: 98% CAS No.: 723-89-7 Molecular Formula: C14H9NO2 Formula Weight: 223.23 Melting Point: 139-142° Beilstein Reference: 164531

Formula: C₁₄H₉NO₂
Boiling Pt: 404 °C (1013 hPa)
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00082681
CAS Number: 723-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR