2-Amino-4-methoxybenzothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxybenzothiazol-2-ylamine

L11358-03 L11358-06 L11358-14
CAAAAL11358-03EA 79.32 CAD
CAAAAL11358-03 CAAAAL11358-06 CAAAAL11358-14
2-Amino-4-methoxybenzothiazole 97%
2-Amino-4-methoxybenzothiazole

Purity: 97% CAS No.: 5464-79-9 Molecular Formula: C8H8N2OS Formula Weight: 180.23 Melting Point: 153-159° Beilstein Reference: 141363

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 335
Melting Pt: 153…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00005792
CAS Number: 5464-79-9
EINECS: 226-763-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR