(Bromomethyl)cyclohexane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11343-06
L11343-14
CAAAAL11343-06EA
47.56
CAD
CAAAAL11343-06
CAAAAL11343-14
(Bromomethyl)cyclohexane 98%
(Bromomethyl)cyclohexane
Purity: 98%
CAS No.: 2550-36-9
Molecular Formula: C7H13Br
Formula Weight: 177.09
Flash Point: 68°(154°F)
Refractive Index: 1.4935
Boiling Point: 99-100°/40mm
Beilstein Reference: 635738
MDL No.: MFCD00001509
Formula:
C₇H₁₃Br Boiling Pt: 99…100 °C/40 mm Density: 1.269 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001509 CAS Number: 2550-36-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...