3-Isothiocyanatobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11322-03
L11322-06
CAAAAL11322-03EA
114.08
CAD
CAAAAL11322-03
CAAAAL11322-06
3-Isothiocyanatobenzoic acid 97%
3-Isothiocyanatobenzoic acid
Purity: 97%
CAS No.: 2131-63-7
Molecular Formula: C8H5NO2S
Formula Weight: 179.20
Odor: Irritating
Melting Point: ca 165° dec.
Beilstein Reference: 2209003
UN No.: 2923
MDL No.: MFCD00041367
Formula:
C₈H₅NO₂S Boiling Pt: 336 °C (1013 hPa) Melting Pt: ∼ 165 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041367 CAS Number: 2131-63-7 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...