(S)-(+)-2-Amino-3-methyl-1-butanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11300-03
L11300-06
L11300-14
CAAAAL11300-03EA
70.47
CAD
CAAAAL11300-03
CAAAAL11300-06
CAAAAL11300-14
(S)-(+)-2-Amino-3-methyl-1-butanol 97%
(S)-(+)-2-Amino-3-methyl-1-butanol
Purity: 97%
CAS No.: 2026-48-4
Molecular Formula: C5H13NO
Formula Weight: 103.17
Storage Temperature: 2°C to 8°C
Flash Point: 78°(172°F)
Refractive Index: 1.4548
Melting Point: 30-35°
Boiling Point: 80-81°/8mm
Beilstein Reference: 1719137
Optical Rotation: +17° (c=10 in ethanol)
MDL No.: MFCD00064296
Formula:
C₅H₁₃NO Boiling Pt: 80…81 °C/8 mm Melting Pt: 30…35 °C Density: 0,926 Flash Pt: 78 °C(172 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064296 CAS Number: 2026-48-4 EINECS: 217-975-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...