trans-1,2-Cyclohexanedicarboxylic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: trans-Cyclohexane-1,2-dicarboxylic acid

L11288-06 L11288-14
CAAAAL11288-06EA 98.75 CAD
CAAAAL11288-06 CAAAAL11288-14
trans-1,2-Cyclohexanedicarboxylic acid 98%
trans-1,2-Cyclohexanedicarboxylic acid

Purity: 98% CAS No.: 2305-32-0 Molecular Formula: C8H12O4 Formula Weight: 172.18 Melting Point: 220-221° Beilstein Reference: 3200609

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 220…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00001464
CAS Number: 2305-32-0
EINECS: 218-975-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR