1,2-Benzenedithiol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzene-1,2-dithiol
L11253-03
L11253-06
L11253-MD
CAAAAL11253-03EA
225.94
CAD
CAAAAL11253-03
CAAAAL11253-06
CAAAAL11253-MD
1,2-Benzenedithiol 97%
1,2-Benzenedithiol
Purity: 97%
CAS No.: 17534-15-5
Molecular Formula: C6H6S2
Formula Weight: 142.24
Storage Temperature: 2°C to 8°C
Flash Point: 103°(217°F)
Refractive Index: 1.6580
Odor: Unpleasant
Melting Point: 21-25°
Boiling Point: 239°
Beilstein Reference: 636154
UN No.: 3335
MDL No.: MFCD00004835
Formula:
C₆H₆S₂ Boiling Pt: 239 °C Melting Pt: 21…25 °C Density: 1.236 Flash Pt: 103 °C(217 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004835 CAS Number: 17534-15-5 EINECS: 241-530-4 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...