2-Chlorothiobenzamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11180-06
L11180-14
CAAAAL11180-06EA
135.25
CAD
CAAAAL11180-06
CAAAAL11180-14
2-Chlorothiobenzamide 97%
2-Chlorothiobenzamide
Purity: 97%
CAS No.: 15717-17-6
Molecular Formula: C7H6ClNS
Formula Weight: 171.65
Melting Point: 63-66°
Beilstein Reference: 508511
Formula:
C₇H₆ClNS Boiling Pt: 316 °C (1013 hPa) Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040955 CAS Number: 15717-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...