5-Methoxy-1-indanone 98%

Supplier: Thermo Scientific Chemicals
Warning

L11165-03 L11165-06
CAAAAL11165-03EA 62.88 CAD
CAAAAL11165-03 CAAAAL11165-06
5-Methoxy-1-indanone 98%
5-Methoxy-1-indanone

Purity: 98% CAS No.: 5111-70-6 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 108-110° Beilstein Reference: 1282137

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 145 - 150 °C (0.2 -rr)
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003789
CAS Number: 5111-70-6
EINECS: 225-838-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR