α,α,α-Trifluoro-o-tolyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11156-03
L11156-06
L11156-14
CAAAAL11156-03EA
40.76
CAD
CAAAAL11156-03
CAAAAL11156-06
CAAAAL11156-14
α,α,α-Trifluoro-o-tolyl isocyanate 97%
α,α,α-Trifluoro-o-tolyl isocyanate
Purity: 97%
CAS No.: 2285-12-3
Molecular Formula: C8H4F3NO
Formula Weight: 187.12
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.4755
Odor: Pungent
Boiling Point: 74-75°/40mm
Beilstein Reference: 977056
UN No.: 2285
MDL No.: MFCD00002008
Formula:
C₈H₄F₃NO Boiling Pt: 74…75 °C/40 mm Density: 1.238 Flash Pt: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002008 CAS Number: 2285-12-3 EINECS: 218-925-5 UN: 3384 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...