1,1,1-Trifluoro-2-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

L11150-06 L11150-14
CAAAAL11150-06EA 103.17 CAD
CAAAAL11150-06 CAAAAL11150-14
1,1,1-Trifluoro-2-propanol 97%
1,1,1-Trifluoro-2-propanol

Purity: 97% CAS No.: 374-01-6 Molecular Formula: C3H5F3O Formula Weight: 114.07 Flash Point: 25°(77°F) Refractive Index: 1.3175 Boiling Point: 75-76° Beilstein Reference: 1734738 UN No.: 1987 MDL No.: MFCD00014403

Formula: C₃H₅F₃O
MW: 114.07
Boiling Pt: 75…76 °C
Melting Pt: –52 °C
Density: 1.259
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014403
CAS Number: 374-01-6
EINECS: 206-773-2
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR