(S)-(-)-1-Phenylethyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11141-03
L11141-06
CAAAAL11141-03EA
99.54
CAD
CAAAAL11141-03
CAAAAL11141-06
(S)-(-)-1-Phenylethyl isocyanate 98%
(S)-(-)-1-Phenylethyl isocyanate
Purity: 98%
CAS No.: 14649-03-7
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5145
Odor: Pungent
Boiling Point: 55-56°/2.5mm
Beilstein Reference: 4230972
Optical Rotation: -10.5° (neat)
UN No.: 2206
MDL No.: MFCD00002036
Formula:
C₉H₉NO Boiling Pt: 55…56 °C/2.5 mm Density: 1.045 Flash Pt: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002036 CAS Number: 14649-03-7 EINECS: 238-698-6 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...