3-Chloro-4-fluorothiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

L11132-03 L11132-MD
CAAAAL11132-03EA 173.8 CAD
CAAAAL11132-03 CAAAAL11132-MD
3-Chloro-4-fluorothiobenzamide 97%
3-Chloro-4-fluorothiobenzamide

Purity: 97% CAS No.: 130560-97-3 Molecular Formula: C7H5ClFNS Formula Weight: 189.64 Melting Point: 129-130° Beilstein Reference: 7757675 UN No.: 2811 MDL No.: MFCD00042478

Formula: C₇H₅ClFNS
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00042478
CAS Number: 130560-97-3
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR