Diallyl disulfide tech. 80% remainder mainly diallyl sulfide and diallyl trisulfide, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11106-14
L11106-22
CAAAAL11106-14EA
96.54
CAD
CAAAAL11106-14
CAAAAL11106-22
Diallyl disulfide tech. 80% remainder mainly diallyl sulfide and diallyl trisulfide, Technical Grade
Diallyl disulphide
Purity: 80%
CAS No.: 2179-57-9
Molecular Formula: C6H10S2
Formula Weight: 146.27
Storage Temperature: 2°C to 8°C
Flash Point: 51°(123°F)
Refractive Index: 1.5410
Odor: Garlic-like
Boiling Point: 180-190°
Beilstein Reference: 1699241
UN No.: 1993
MDL No.: MFCD00008656
Formula:
C₆H₁₀S₂ Boiling Pt: 180…190 °C Density: 1.008 Flash Pt: 51 °C (123 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008656 CAS Number: 2179-57-9 EINECS: 218-548-6 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...