2-Hydroxybenzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone

L11100-06 L11100-14
CAAAAL11100-06EA 97.64 CAD
CAAAAL11100-06 CAAAAL11100-14
2-Hydroxybenzophenone 99%
2-Hydroxybenzophenone

Purity: 99% CAS No.: 117-99-7 Molecular Formula: C13H10O2 Formula Weight: 198.22 Melting Point: 38-40° Boiling Point: 171-173°/12mm MDL No.: MFCD00002216

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 171…173 °C (12 mmHg)
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00002216
CAS Number: 117-99-7
EINECS: 204-226-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR