2,4-Dichlorophenyl isocyanate 96%

Supplier: Thermo Scientific Chemicals
Danger

L11089-03 L11089-06 L11089-14
CAAAAL11089-03EA 27.81 CAD
CAAAAL11089-03 CAAAAL11089-06 CAAAAL11089-14
2,4-Dichlorophenyl isocyanate 96%
2,4-Dichlorophenyl isocyanate

Purity: 96% CAS No.: 2612-57-9 Molecular Formula: C7H3Cl2NO Formula Weight: 188.01 Odor: Irritating Melting Point: 57-61° Boiling Point: 80°/1mm Beilstein Reference: 2090780 UN No.: 2250 MDL No.: MFCD00002001

Formula: C₇H₃Cl₂NO
Boiling Pt: 80 °C/1 mm
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00002001
CAS Number: 2612-57-9
EINECS: 220-040-4
UN: 2250
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR