2-Cyclopropylethanol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11080-03
L11080-06
L11080-MD
CAAAAL11080-03EA
180.12
CAD
CAAAAL11080-03
CAAAAL11080-06
CAAAAL11080-MD
2-Cyclopropylethanol 96%
2-Cyclopropylethanol
Purity: 96%
CAS No.: 2566-44-1
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 47°(116°F)
Refractive Index: 1.4355
Boiling Point: 137-138°
Beilstein Reference: 2036028
UN No.: 1987
MDL No.: MFCD00040762
Formula:
C₅H₁₀O Boiling Pt: 137…138 °C Flash Pt: 47 °C(116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040762 CAS Number: 2566-44-1 UN: 1987 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...