1-Bromo-2,6-dichlorobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

L11062-06 L11062-14
CAAAAL11062-06EA 55.77 CAD
CAAAAL11062-06 CAAAAL11062-14
1-Bromo-2,6-dichlorobenzene 97%
1-Bromo-2,6-dichlorobenzene

Purity: 97% CAS No.: 19393-92-1 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Flash Point: 100°(212°F) Melting Point: 59-62° Boiling Point: 241-243° Beilstein Reference: 1681054 MDL No.: MFCD00000574

Formula: C₆H₃BrCl₂
Boiling Pt: 241…243 °C
Melting Pt: 59…62 °C
Density: 2.077 g/cm³ (20 °C)
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000574
CAS Number: 19393-92-1
EINECS: 243-018-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR