Methyl isothiocyanatoacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11046-03
L11046-06
CAAAAL11046-03EA
64.31
CAD
CAAAAL11046-03
CAAAAL11046-06
Methyl isothiocyanatoacetate 98%
Methyl isothiocyanatoacetate
Purity: 98%
CAS No.: 21055-37-8
Molecular Formula: C4H5NO2S
Formula Weight: 131.15
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5200
Odor: Irritating
Boiling Point: 137-138°/35mm
Beilstein Reference: 2074764
UN No.: 2922
MDL No.: MFCD00041119
Formula:
C₄H₅NO₂S Boiling Pt: 137…138 °C/35 mm Density: 1.210 Storage Temperature: Refrigerator |
MDL Number:
MFCD00041119 CAS Number: 21055-37-8 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...