(R)-(-)-2-Amino-1-propanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11030-03
L11030-06
L11030-14
CAAAAL11030-03EA
61.15
CAD
CAAAAL11030-03
CAAAAL11030-06
CAAAAL11030-14
(R)-(-)-2-Amino-1-propanol 98%
(R)-(-)-2-Amino-1-propanol
Purity: 98%
CAS No.: 35320-23-1
Molecular Formula: C3H9NO
Formula Weight: 75.11
Flash Point: 83°(181°F)
Refractive Index: 1.4493
Boiling Point: 174-176°
Beilstein Reference: 1718866
Optical Rotation: -22° (c=2 in ethanol)
UN No.: 2735
MDL No.: MFCD00064413
Formula:
C₃H₉NO Boiling Pt: 174…176 °C Density: 0.965 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064413 CAS Number: 35320-23-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...