Diallyl sulfide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Allyl sulphide, Allyl sulfide
L10971-14
L10971-22
CAAAAL10971-14EA
52.14
CAD
CAAAAL10971-14
CAAAAL10971-22
Diallyl sulfide 98%
Diallyl sulphide
Purity: 98%
CAS No.: 592-88-1
Molecular Formula: C6H10S
Formula Weight: 114.21
Flash Point: 23°(73°F)
Refractive Index: 1.4895
Odor: Unpleasant
Melting Point: -83°
Boiling Point: 137-138°
Beilstein Reference: 1736016
Merck Reference: 14,297
UN No.: 1993
MDL No.: MFCD00008658
Formula:
C₆H₁₀S MW: 114.21 g/mol Boiling Pt: 137…138 °C Melting Pt: –83 °C Density: 0.889 Flash Pt: 23 °C (73 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008658 CAS Number: 592-88-1 EINECS: 209-775-1 UN: 1993 ADR: 3,III Merck Index: 14,297 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...