(α,α,α-Trifluoro-3-tolyl)acetic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-(Trifluoromethyl)phenyl)acetic acid, 3-(Trifluoromethyl)phenylacetic acid

L10960-09 L10960-18
CAAAAL10960-09EA 141.25 CAD
CAAAAL10960-09 CAAAAL10960-18
(α,α,α-Trifluoro-3-tolyl)acetic acid 96%
(α,α,α-Trifluoro-3-tolyl)acetic acid

Purity: 96% CAS No.: 351-35-9 Molecular Formula: C9H7F3O2 Formula Weight: 204.15 Melting Point: 77-78° Beilstein Reference: 2213223

Formula: (C₆H₄(CF₃))CH₂COOH
MW: 204.15 g/mol
Melting Pt: 77…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004339
CAS Number: 351-35-9
EINECS: 206-511-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR