4-Methoxythiobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxybenzothioamide

L10945-03 L10945-06
CAAAAL10945-03EA 58.78 CAD
CAAAAL10945-03 CAAAAL10945-06
4-Methoxythiobenzamide 98%
4-Methoxythiobenzamide

Purity: 98% CAS No.: 2362-64-3 Molecular Formula: C8H9NOS Formula Weight: 167.23 Melting Point: 145-147° Beilstein Reference: 1072444

Formula: C₈H₉NOS
MW: 167.23
Boiling Pt: 282
Melting Pt: 155
Storage Temperature: Ambient
MDL Number: MFCD00040993
CAS Number: 2362-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR