Ethyl acetate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid ethyl ester

L10925-0F L10925-AK L10925-AU
CAAAAL10925-0FEA 116.76 CAD
CAAAAL10925-0F CAAAAL10925-AK CAAAAL10925-AU
Ethyl acetate 99%
Ethyl acetate

Purity: 99% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Refractive Index: 1.3720 Odor: Sweet Melting Point: -84° Boiling Point: 76-77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.902
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 14,3757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR