3-Chloro-4-fluorophenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10891-06
L10891-14
CAAAAL10891-06EA
121.34
CAD
CAAAAL10891-06
CAAAAL10891-14
3-Chloro-4-fluorophenyl isocyanate 98%
3-Chloro-4-fluorophenyl isocyanate
Purity: 98%
CAS No.: 50529-33-4
Molecular Formula: C7H3ClFNO
Formula Weight: 171.56
Storage Temperature: 2°C to 8°C
Flash Point: 87°(188°F)
Refractive Index: 1.5410
Boiling Point: 44°/17mm
Beilstein Reference: 973324
UN No.: 2206
MDL No.: MFCD00037037
Formula:
C₇H₃ClFNO Boiling Pt: 44 °C/17 mm Density: 1.377 Flash Pt: 87 °C (188 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037037 CAS Number: 50529-33-4 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...