Methyl-3,5-diaminobenzoate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3,5-Diaminobenzoic acid methyl ester, Methyl 3,5-diaminobenzoate
L10848-09
L10848-18
CAAAAL10848-09EA
115.34
CAD
CAAAAL10848-09
CAAAAL10848-18
Methyl-3,5-diaminobenzoate 99%
Methyl 3,5-diaminobenzoate
Purity: 99%
CAS No.: 1949-55-9
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Odor: Odorless
Melting Point: 131-134°
Beilstein Reference: 2363309
MDL No.: MFCD00041433
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Boiling Pt: 319 Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041433 CAS Number: 1949-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...