2-Methoxyethyl isothiocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10839-03
L10839-06
CAAAAL10839-03EA
43.61
CAD
CAAAAL10839-03
CAAAAL10839-06
2-Methoxyethyl isothiocyanate 98+%
2-Methoxyethyl isothiocyanate
Purity: 98+%
CAS No.: 38663-85-3
Molecular Formula: CH3O(CH2)2NCS
Formula Weight: 117.17
Storage Temperature: 2°C to 8°C
Flash Point: 66°(150°F)
Refractive Index: 1.5120
Boiling Point: 88-92°/24mm
Beilstein Reference: 773990
UN No.: 2922
MDL No.: MFCD00041126
Formula:
C₄H₇NOS MW: 117.17 g/mol Boiling Pt: 88…92 °C (24 mmHg) Density: 1.080 Flash Pt: 66 °C (150 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00041126 CAS Number: 38663-85-3 EINECS: 254-070-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...