4-Bromophenyl isocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10834-06
L10834-14
CAAAAL10834-06EA
75.21
CAD
CAAAAL10834-06
CAAAAL10834-14
4-Bromophenyl isocyanate 99%
4-Bromophenyl isocyanate
Purity: 99%
CAS No.: 2493-02-9
Molecular Formula: C7H4BrNO
Formula Weight: 198.03
Storage Temperature: 2°C to 8°C
Flash Point: 109°(228°F)
Odor: Pungent
Melting Point: 39-42°
Boiling Point: 157-158°/14mm
Beilstein Reference: 508203
Merck Reference: 14,1430
UN No.: 2811
MDL No.: MFCD00002022
Formula:
C₇H₄BrNO Boiling Pt: 157…158 °C/14 mm Melting Pt: 39…42 °C Flash Pt: 109 °C(228 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002022 CAS Number: 2493-02-9 EINECS: 219-662-9 UN: 3335 ADR: 6.1,II Merck Index: 14,1430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...