3-Hydroxy-1-methylpiperidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Methylpiperidin-3-ol, N-Methyl-3-piperidinol
L10829-06
L10829-14
CAAAAL10829-06EA
43.13
CAD
CAAAAL10829-06
CAAAAL10829-14
3-Hydroxy-1-methylpiperidine 98%
3-Hydroxy-1-methylpiperidine
Purity: 98%
CAS No.: 3554-74-3
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 70°(158°F)
Refractive Index: 1.4740
Boiling Point: 76-78°/11mm
Beilstein Reference: 103017
MDL No.: MFCD00006495
Formula:
C₆H₁₃NO MW: 115.18 g/mol Boiling Pt: 76…78 °C (11 mmHg) Density: 0.999 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006495 CAS Number: 3554-74-3 EINECS: 222-609-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...