1,1-Dibromo-3,3,3-trifluoroacetone 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one

L10826-06 L10826-14
CAAAAL10826-06EA 131.61 CAD
CAAAAL10826-06 CAAAAL10826-14
1,1-Dibromo-3,3,3-trifluoroacetone 95%
1,1-Dibromo-3,3,3-trifluoroacetone

Purity: 95% CAS No.: 431-67-4 Molecular Formula: C3HBr2F3O Formula Weight: 269.85 Storage Temperature: 2°C to 8°C Refractive Index: 1.4305 Odor: Pungent Boiling Point: 111-113° Beilstein Reference: 636645 UN No.: 3265 MDL No.: MFCD00041362

Formula: Br₂HCC(O)CF₃
Boiling Pt: 111…113 °C
Density: 2.188 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00041362
CAS Number: 431-67-4
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR