Propiolaldehyde diethyl acetal 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne
L10767-03
L10767-06
L10767-14
CAAAAL10767-03EA
76.79
CAD
CAAAAL10767-03
CAAAAL10767-06
CAAAAL10767-14
Propiolaldehyde diethyl acetal 97%
Propiolaldehyde diethyl acetal
Purity: 97%
CAS No.: 10160-87-9
Molecular Formula: HC≡CCH(OCH2CH3)2
Formula Weight: 128.17
Storage Temperature: 2°C to 8°C
Flash Point: 32°(89°F)
Refractive Index: 1.4120
Boiling Point: 138-139°
Beilstein Reference: 1701566
UN No.: 1993
MDL No.: MFCD00009237
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 138…139 °C Density: 0.894 Flash Pt: 32 °C (89 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009237 CAS Number: 10160-87-9 EINECS: 233-430-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...