2,6-Diisopropylaniline 90+%, Technical Grade
Supplier: Thermo Scientific Chemicals
L10761-22
L10761-36
CAAAAL10761-22EA
62.88
CAD
CAAAAL10761-22
CAAAAL10761-36
2,6-Diisopropylaniline 90+%, Technical Grade
2,6-Diisopropylaniline
Purity: 90+%
CAS No.: 24544-04-5
Molecular Formula: C12H19N
Formula Weight: 177.29
Flash Point: 125°(257°F)
Refractive Index: 1.5330
Melting Point: -54°
Boiling Point: 254-257°
Beilstein Reference: 2208763
MDL No.: MFCD00008887
Formula:
C₁₂H₁₉N Boiling Pt: 254…257 °C Melting Pt: –54 °C Density: 0.940 Flash Pt: 125 °C(257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008887 CAS Number: 24544-04-5 EINECS: 246-305-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...