Cyclohexylethene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10756-06
L10756-14
CAAAAL10756-06EA
107.12
CAD
CAAAAL10756-06
CAAAAL10756-14
Cyclohexylethene 97%
Cyclohexylethene
Purity: 97%
CAS No.: 695-12-5
Molecular Formula: C8H14
Formula Weight: 110.20
Flash Point: 22°(71°F)
Refractive Index: 1.4465
Boiling Point: 127-128°
Beilstein Reference: 1304663
UN No.: 3295
MDL No.: MFCD00001514
Formula:
C₈H₁₄ Boiling Pt: 127…128 °C Density: 0.805 Flash Pt: 22 °C (71 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001514 CAS Number: 695-12-5 EINECS: 211-779-3 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...