2-Chloro-4-(trifluoromethyl)pyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10713-03
L10713-06
CAAAAL10713-03EA
118.97
CAD
CAAAAL10713-03
CAAAAL10713-06
CAAAAL10713-MD
2-Chloro-4-(trifluoromethyl)pyridine 98+%
2-Chloro-4-(trifluoromethyl)pyridine
Purity: 98+%
CAS No.: 81565-18-6
Molecular Formula: C6H3ClF3N
Formula Weight: 181.55
Flash Point: 40°(104°F)
Refractive Index: 1.4490
Boiling Point: 145°
Beilstein Reference: 4246276
UN No.: 1992
MDL No.: MFCD00042224
Formula:
C₆H₃ClF₃N Boiling Pt: 145 °C Melting Pt: –1 °C Density: 1.411 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042224 CAS Number: 81565-18-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...