(±)-2,4-Difluoromandelic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-2,4-Difluorophenylglycolic acid

L10708-03 L10708-06
CAAAAL10708-03EA 107.6 CAD
CAAAAL10708-03 CAAAAL10708-06
(±)-2,4-Difluoromandelic acid 97%
(±)-2,4-Difluoromandelic acid

Purity: 97% CAS No.: 132741-30-1 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 122-124° MDL No.: MFCD00061296

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00061296
CAS Number: 132741-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR