trans-5-Decene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10689-03
L10689-06
CAAAAL10689-03EA
63.83
CAD
CAAAAL10689-03
CAAAAL10689-06
trans-5-Decene 97%
trans-5-Decene
Purity: 97%
CAS No.: 7433-56-9
Molecular Formula: C10H20
Formula Weight: 140.27
Flash Point: 46°(114°F)
Refractive Index: 1.4240
Boiling Point: 171-172°
Beilstein Reference: 1719487
UN No.: 3295
MDL No.: MFCD00009508
Formula:
C₁₀H₂₀ Boiling Pt: 171…172 °C Melting Pt: –73 °C Density: 0.740 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009508 CAS Number: 7433-56-9 EINECS: 231-080-7 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...