N-Isopropylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10646-09
L10646-18
CAAAAL10646-09EA
78.84
CAD
CAAAAL10646-09
CAAAAL10646-18
N-Isopropylaniline 98%
N-Isopropylaniline
Purity: 98%
CAS No.: 768-52-5
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 88°(190°F)
Refractive Index: 1.5385
Melting Point: -32°
Boiling Point: 203-204°
Beilstein Reference: 2205871
UN No.: 2810
MDL No.: MFCD00026347
Formula:
C₆H₅NHCH(CH₃)₂ Boiling Pt: 203…204 °C Melting Pt: –32 °C Density: 0.938 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026347 CAS Number: 768-52-5 EINECS: 212-196-7 UN: 2810 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...