2-Isopropylphenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Isopropylphenylisocyanate
L10618-06
L10618-14
CAAAAL10618-06EA
92.59
CAD
CAAAAL10618-06
CAAAAL10618-14
2-Isopropylphenyl isocyanate 97%
2-Isopropylphenyl isocyanate
Purity: 97%
CAS No.: 56309-56-9
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Flash Point: 87°(188°F)
Refractive Index: 1.5230
Boiling Point: 66-67°/5mm
Beilstein Reference: 3588928
UN No.: 2206
MDL No.: MFCD00013850
Formula:
C₁₀H₁₁NO Boiling Pt: 66…67 °C/5 mm Density: 1.019 Flash Pt: 87 °C (188 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013850 CAS Number: 56309-56-9 EINECS: 260-105-4 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...