2,6-Diisopropylphenyl isocyanate 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10613-03
L10613-06
L10613-14
CAAAAL10613-03EA
36.18
CAD
CAAAAL10613-03
CAAAAL10613-06
CAAAAL10613-14
2,6-Diisopropylphenyl isocyanate 94%
2,6-Diisopropylphenyl isocyanate
Purity: 94%
CAS No.: 28178-42-9
Molecular Formula: C13H17NO
Formula Weight: 203.29
Storage Temperature: 2°C to 8°C
Flash Point: 107°(224°F)
Refractive Index: 1.5180
Boiling Point: 116-117°/10mm
Beilstein Reference: 2645721
UN No.: 2206
MDL No.: MFCD00008882
Formula:
C₁₃H₁₇NO Boiling Pt: 116…117 °C/10 mm Density: 0.951 Flash Pt: 107 °C (224 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008882 CAS Number: 28178-42-9 EINECS: 248-885-4 UN: 2927 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...